Tisk…

3-(Bromomethyl)-2-nitropyridine

Dodavatel: Apollo Scientific

OR200119-250MG
APOSOR200119-250MGEA 13460 CZK
APOSOR200119-250MG
3-(Bromomethyl)-2-nitropyridine
3-(Bromomethyl)-2-nitropyridine
Vzorec: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14584511
CAS číslo: 15936-06-8
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR