Ethyl 2-amino-5-iodothiazole-4-carboxylate
Dodavatel: Apollo Scientific
OR200115-1G
OR200115-250MG
OR200115-500MG
APOSOR200115-1GEA
6350
CZK
APOSOR200115-1G
APOSOR200115-250MG
APOSOR200115-500MG
Ethyl 2-amino-5-iodothiazole-4-carboxylate
Ethyl 2-amino-5-iodothiazole-4-carboxylate
Vzorec:
C₆H₇IN₂O₂S MW: 298,101 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11975710 CAS číslo: 860646-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...