3-Chloro-5-methylbenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Chloro-m-toluic acid, 3-Carboxy-5-chlorotoluene
OR19520-10G
OR19520-25G
APOSOR19520-10GEA
2610
CZK
APOSOR19520-10G
APOSOR19520-25G
3-Chloro-5-methylbenzoic acid 95%
3-Chloro-5-methylbenzoic acid
Vzorec:
C₈H₇ClO₂ MW: 170.593 g/mol Bod tání: 181…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD11007823 CAS číslo: 56961-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...