4-Amino-3-ethoxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Carboxy-2-ethoxyaniline, 2-Amino-5-carboxyphenetole
OR18857-1G
OR18857-250MG
APOSOR18857-1GEA
3970
CZK
APOSOR18857-1G
APOSOR18857-250MG
4-Amino-3-ethoxybenzoic acid
4-Amino-3-ethoxybenzoic acid
Vzorec:
C₉H₁₁NO₃ MW: 181,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12168720 CAS číslo: 367501-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...