Tisk…

Methyl 6-chloropyrimidine-4-carboxylate

Dodavatel: Apollo Scientific

OR18854-1G OR18854-5G
APOSOR18854-1GEA 734 CZK
APOSOR18854-1G APOSOR18854-5G
Methyl 6-chloropyrimidine-4-carboxylate
Methyl 6-chloropyrimidine-4-carboxylate
Vzorec: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08689467
CAS číslo: 6627-22-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR