Tisk…

(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4R)-3-(tert-Butoxycarbonyl)-4-methyl-1,2,3-oxathiazolidine 2,2-dioxide, (R)-3-Boc-4-methyl-1,2,3-oxathiazolidine 2,2-Dioxide, tert-Butyl (4R)-4-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide

OR18837-1G OR18837-5G OR18837-25G
APOSOR18837-1GEA 1450 CZK
APOSOR18837-1G APOSOR18837-5G APOSOR18837-25G
(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected 95+%
(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected
Vzorec: C₈H₁₅NO₅S
MW: 237,273 g/mol
Bod tání: 121…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864994
CAS číslo: 454248-53-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR