(Bicyclo[1.1.1]pent-1-yl)methanol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Bicyclo[1.1.1]pentan-1-ylmethanol, 1-(Hydroxymethyl)bicyclo[1.1.1]pentane
OR18811-250MG
OR18811-100MG
OR18811-25MG
APOSOR18811-250MGEA
21730
CZK
APOSOR18811-250MG
APOSOR18811-100MG
APOSOR18811-25MG
(Bicyclo[1.1.1]pent-1-yl)methanol
(Bicyclo[1.1.1]pent-1-yl)methanol
Vzorec:
C₆H₁₀O MW: 98,143 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28040494 CAS číslo: 22287-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...