Tisk…

2-Amino-4-chloro-1,3-thiazole-5-carboxaldehyde, 2-BOC protected

Dodavatel: Apollo Scientific
Varování

OR18806-1G OR18806-250MG OR18806-5G
APOSOR18806-1GEA 9390 CZK
APOSOR18806-1G APOSOR18806-250MG APOSOR18806-5G
2-Amino-4-chloro-1,3-thiazole-5-carboxaldehyde, 2-BOC protected
2-Amino-4-chloro-1,3-thiazole-5-carboxaldehyde, 2-BOC protected
Vzorec: C₉H₁₁ClN₂O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD18836516
CAS číslo: 388594-31-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR