Tisk…

Methyl 3-amino-2,6-dichloroisonicotinate 95%

Dodavatel: Apollo Scientific

OR18588-1G OR18588-250MG
APOSOR18588-1GEA 0 CZK
APOSOR18588-1G APOSOR18588-250MG
Methyl 3-amino-2,6-dichloroisonicotinate 95%
Methyl 3-amino-2,6-dichloroisonicotinate
Vzorec: C₇H₆Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07437844
CAS číslo: 883107-62-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR