Tisk…

1,6-Dimethyl-3-iodo-1H-indazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Iodo-1,6-Dimethyl-1H-indazole

OR18423-1G OR18423-250MG
APOSOR18423-1GEA 5790 CZK
APOSOR18423-1G APOSOR18423-250MG
1,6-Dimethyl-3-iodo-1H-indazole
1,6-Dimethyl-3-iodo-1H-indazole
Vzorec: C₉H₉IN₂
MW: 272,086 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16628058
CAS číslo: 1257535-50-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR