Tisk…

4-(Piperidin-1-yl)benzene-1,2-diamine

Dodavatel: Apollo Scientific

OR183525-1G OR183525-250MG
APOSOR183525-1GEA 15710 CZK
APOSOR183525-1G APOSOR183525-250MG
4-(Piperidin-1-yl)benzene-1,2-diamine
4-(Piperidin-1-yl)benzene-1,2-diamine
Vzorec: C₁₁H₁₇N₃
MW: 191,273 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848949
CAS číslo: 155198-10-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR