Tisk…

2-Chloro-1-isopropyl-1H-imidazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-1-(prop-2-yl)-1H-imidazole

OR17928-1G OR17928-250MG
APOSOR17928-1GEA 14980 CZK
APOSOR17928-1G APOSOR17928-250MG
2-Chloro-1-isopropyl-1H-imidazole
2-Chloro-1-isopropyl-1H-imidazole
Vzorec: C₆H₉ClN₂
MW: 144,602 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699127
CAS číslo: 1053655-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR