Tisk…

4-Bromo-3-(methylsulphonyl)aniline 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Amino-2-bromophenyl methyl sulphone

OR17676-1G OR17676-250MG OR17676-5G
APOSOR17676-1GEA 13230 CZK
APOSOR17676-1G APOSOR17676-250MG APOSOR17676-5G
4-Bromo-3-(methylsulphonyl)aniline 95%
4-Bromo-3-(methylsulphonyl)aniline
Vzorec: C₇H₈BrNO₂S
MW: 250,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602552
CAS číslo: 1375473-63-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR