Tisk…

5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

OR17548-1G OR17548-250MG
APOSOR17548-1GEA 20770 CZK
APOSOR17548-1G APOSOR17548-250MG
5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole
5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole
Vzorec: C₆H₁₀BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD08449281
CAS číslo: 1017788-72-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR