3-Bromopyridine N-oxide hydrochloride 97%
Dodavatel: Apollo Scientific
OR17508-1G
OR17508-25G
OR17508-5G
APOSOR17508-1GEA
2570
CZK
APOSOR17508-1G
APOSOR17508-25G
APOSOR17508-5G
3-Bromopyridine N-oxide hydrochloride 97%
3-Bromopyridine N-oxide hydrochloride
Vzorec:
C₅H₄BrNO·ClH MW: 210,456 g/mol Bod tání: 180…181 °C Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01940270 CAS číslo: 63785-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...