2-Bromo-6-hydroxybenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-2-carboxyphenol, 6-Bromosalicylic acid
OR17501-1G
OR17501-250MG
APOSOR17501-1GEA
8270
CZK
APOSOR17501-1G
APOSOR17501-250MG
2-Bromo-6-hydroxybenzoic acid 98%
2-Bromo-6-hydroxybenzoic acid
Vzorec:
C₇H₅BrO₃ MW: 217,017 g/mol Bod tání: 164…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD13193059 CAS číslo: 38876-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...