3-Hydroxy-4-methylbenzeneboronic acid
Dodavatel: Apollo Scientific
Synonyms:
5-Borono-2-methylphenol, (3-Hydroxy-4-methylphenyl)boronic acid, 3-Hydroxy-4-methylphenylboronic acid, 5-Borono-o-cresol, 4-Borono-2-hydroxytoluene
OR17103-1G
OR17103-5G
APOSOR17103-1GEA
12730
CZK
APOSOR17103-1G
APOSOR17103-5G
3-Hydroxy-4-methylbenzeneboronic acid
3-Hydroxy-4-methylbenzeneboronic acid
Vzorec:
C₇H₉BO₃ MW: 151,956 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01074652 CAS číslo: 216019-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...