Tisk…

3-Amino-2-cyanopropanoic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-2-carboxypropionitrile, 3-Amino-2-cyanopropionic acid

OR17008-1G OR17008-5G
APOSOR17008-1GEA 6250 CZK
APOSOR17008-1G APOSOR17008-5G
3-Amino-2-cyanopropanoic acid 97%
3-Amino-2-cyanopropanoic acid
Vzorec: C₄H₆N₂O₂
MW: 114,103 g/mol
Bod tání: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD10565611
CAS číslo: 1211318-03-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR