Tisk…

2-Amino-4-methylnicotinonitrile

Dodavatel: Apollo Scientific
Varování

OR17000-1G OR17000-5G
APOSOR17000-1GEA 2980 CZK
APOSOR17000-1G APOSOR17000-5G
2-Amino-4-methylnicotinonitrile
2-Amino-4-methylnicotinonitrile
Vzorec: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD09030794
CAS číslo: 71493-76-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR