1-(3-Aminoprop-1-yl)piperazine, 1-BOC protected 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl [3-(piperazin-1-yl)prop-1-yl]carbamate, 1-{3-[(tert-Butoxycarbonyl)amino]prop-1-yl}piperazine, tert-Butyl (3-(piperazin-1-yl)propyl)carbamate
OR16694-1G
OR16694-5G
APOSOR16694-1GEA
4860
CZK
APOSOR16694-1G
APOSOR16694-5G
1-(3-Aminoprop-1-yl)piperazine, 1-BOC protected 97%
1-(3-Aminoprop-1-yl)piperazine, 1-BOC protected
Vzorec:
C₁₂H₂₅N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06808577 CAS číslo: 874831-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...