Tisk…

3-Methyl-2-nitrobenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α-Bromo-2-nitro-m-xylene, 1-(Bromomethyl)-3-methyl-2-nitrobenzene

OR16688-1G OR16688-5G
APOSOR16688-1GEA 1210 CZK
APOSOR16688-1G APOSOR16688-5G
3-Methyl-2-nitrobenzyl bromide 97%
3-Methyl-2-nitrobenzyl bromide
Vzorec: C₈H₈BrNO₂
MW: 230.059 g/mol
Bod tání: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD08276004
CAS číslo: 55324-02-2
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR