Tisk…

β,β-Dimethylphenethylamine

Dodavatel: Apollo Scientific
Nebezpečí

OR16558-1G OR16558-5G OR16558-250MG
APOSOR16558-1GEA 6050 CZK
APOSOR16558-1G APOSOR16558-5G APOSOR16558-250MG
β,β-Dimethylphenethylamine
β,β-Dimethylphenethylamine
Vzorec: C₁₀H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD06213339
CAS číslo: 21404-88-6
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR