4-[(Methylamino)methyl]-4-phenyltetrahydro-2H-pyran
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
N-methyl-1-(4-phenyltetrahydro-2H-pyran-4-yl)methylamine
OR16552-1G
OR16552-250MG
APOSOR16552-1GEA
6220
CZK
APOSOR16552-1G
APOSOR16552-250MG
4-[(Methylamino)methyl]-4-phenyltetrahydro-2H-pyran
4-[(Methylamino)methyl]-4-phenyltetrahydro-2H-pyran
Vzorec:
C₁₃H₁₉NO Storage Temperature: Ambient |
MDL Number:
MFCD11841073 CAS číslo: 958443-30-6 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...