2-(Methylsulphonyl)aniline 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Aminophenyl methyl sulphone
OR16499-1G
OR16499-25G
OR16499-5G
APOSOR16499-1GEA
1160
CZK
APOSOR16499-1G
APOSOR16499-25G
APOSOR16499-5G
2-(Methylsulphonyl)aniline 99%
2-(Methylsulphonyl)aniline
Vzorec:
C₇H₉NO₂S MW: 171,217 g/mol Bod tání: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD08444390 CAS číslo: 2987-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...