Tisk…

Ethyl (2E)-pent-2-enoate, tech

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl trans-pent-2-enoate, (E)-Ethyl pent-2-enoate

OR16435-25G OR16435-5G OR16435-1G
APOSOR16435-25GEA 12400 CZK
APOSOR16435-25G APOSOR16435-5G APOSOR16435-1G
Ethyl (2E)-pent-2-enoate, tech
Ethyl (2E)-pent-2-enoate
Vzorec: C₇H₁₂O₂
MW: 128,169 g/mol
Hustota: 0,9086 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD06204561
CAS číslo: 24410-84-2
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR