Dimethyl 4-aminoisophthalate
Dodavatel: Apollo Scientific
|
Varování
|
OR16406-1G
OR16406-250MG
OR16406-5G
APOSOR16406-1GEA
2690
CZK
APOSOR16406-1G
APOSOR16406-250MG
APOSOR16406-5G
Dimethyl 4-aminoisophthalate
Dimethyl 4-aminoisophthalate
Vzorec:
C₁₀H₁₁NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD12674801 CAS číslo: 63746-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...