Tisk…

3-Amino-5-azaindole dihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 1H-Pyrrolo[3,2-c]pyridin-3-amine dihydrochloride, 3-Amino-1H-pyrrolo[3,2-c]pyridine dihydrochloride

OR15771-1G OR15771-500MG
APOSOR15771-1GEA 27910 CZK
APOSOR15771-1G APOSOR15771-500MG
3-Amino-5-azaindole dihydrochloride
3-Amino-5-azaindole dihydrochloride
Vzorec: C₇H₇N₃·2ClH
MW: 206,072 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16628055
CAS číslo: 1257535-49-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR