Tisk…

3-Amino-6-azaindole dihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-1H-pyrrolo[2,3-c]pyridine dihydrochloride, 1H-Pyrrolo[2,3-c]pyridin-3-amine dihydrochloride

OR15770-1G OR15770-250MG
APOSOR15770-1GEA 12900 CZK
APOSOR15770-1G APOSOR15770-250MG
3-Amino-6-azaindole dihydrochloride
3-Amino-6-azaindole dihydrochloride
Vzorec: C₇H₇N₃·2ClH
MW: 206,072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16628054
CAS číslo: 1257535-55-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR