Tisk…

trans-4-Bromo-3-hydroxypiperidine, N-BOC protected

Dodavatel: Apollo Scientific
Varování

OR15690-1G OR15690-5G
APOSOR15690-1GEA 6550 CZK
APOSOR15690-1G APOSOR15690-5G
trans-4-Bromo-3-hydroxypiperidine, N-BOC protected
trans-4-Bromo-3-hydroxypiperidine, N-BOC protected
Vzorec: C₁₀H₁₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD04972247
CAS číslo: 936250-36-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR