Tisk…

Methyl 5-bromo-2-methyl-1,3-thiazole-4-carboxylate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-4-(methoxycarbonyl)-2-methyl-1,3-thiazole, Methyl 5-bromo-2-methylthiazole-4-carboxylate

OR15542-1G OR15542-250MG OR15542-5G
APOSOR15542-1GEA 2150 CZK
APOSOR15542-1G APOSOR15542-250MG APOSOR15542-5G
Methyl 5-bromo-2-methyl-1,3-thiazole-4-carboxylate 98%
Methyl 5-bromo-2-methyl-1,3-thiazole-4-carboxylate
Vzorec: C₆H₆BrNO₂S
MW: 236,086 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09265508
CAS číslo: 899897-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR