6-(4-Methylphenyl)-1,3,5-triazine-2,4-diamine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(4,6-Diamino-1,3,5-triazin-2-yl)toluene, 2,4-Diamino-6-(4-methylphenyl)-1,3,5-triazine
OR1498-10G
OR1498-1G
OR1498-5G
APOSOR1498-10GEA
7050
CZK
APOSOR1498-10G
APOSOR1498-1G
APOSOR1498-5G
6-(4-Methylphenyl)-1,3,5-triazine-2,4-diamine
6-(4-Methylphenyl)-1,3,5-triazine-2,4-diamine
Vzorec:
C₁₀H₁₁N₅ MW: 201,228 g/mol Bod tání: 232…234 °C Storage Temperature: Ambient |
MDL Number:
MFCD00269392 CAS číslo: 19338-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...