Tisk…

5-Methylthien-2-yl isocyanate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Isocyanato-5-methylthiophene

OR14873-1G OR14873-250MG OR14873-500MG
APOSOR14873-1GEA 5790 CZK
APOSOR14873-1G APOSOR14873-250MG APOSOR14873-500MG
5-Methylthien-2-yl isocyanate 97%
5-Methylthien-2-yl isocyanate

May form small amount of sediment

Vzorec: C₆H₅NOS
Bod varu: 94 °C (53 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09817547
CAS číslo: 76536-99-7
UN: 2206
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR