Tisk…

(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-4-Benzylthiazolidine-2-thione, (4S)-4-(Phenylmethyl)-2-thioxo-1,3-thiazolidine, (4S)-4-Benzyl-2-thioxo-1,3-thiazolidine

OR14521-1G OR14521-10G OR14521-5G
APOSOR14521-1GEA 1620 CZK
APOSOR14521-1G APOSOR14521-10G APOSOR14521-5G
(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%
(4S)-4-Benzyl-1,3-thiazolidine-2-thione
Vzorec: C₁₀H₁₁NS₂
MW: 209,331 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06658216
CAS číslo: 171877-39-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR