Tisk…

1-(Furo[3,2-c]pyridin-4-yl)homopiperazine

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(1,4-Diazepan-1-yl)furo[3,2-c]pyridine, 1-(Furo[3,2-c]pyridin-4-yl)-1,4-diazepane

OR14500-1G OR14500-250MG
APOSOR14500-1GEA 9690 CZK
APOSOR14500-1G APOSOR14500-250MG
1-(Furo[3,2-c]pyridin-4-yl)homopiperazine
1-(Furo[3,2-c]pyridin-4-yl)homopiperazine
Vzorec: C₁₂H₁₅N₃O
MW: 217,267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658987
CAS číslo: 845885-87-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR