Tisk…

tert-Butyl (piperazin-1-yl)acetate

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 2-(piperazin-1-yl)acetate, 1-[(tert-Butoxycarbonyl)methyl]piperazine, (Piperazin-1-yl)acetic acid tert-butyl ester

OR14391-1G OR14391-250MG
APOSOR14391-1GEA 1870 CZK
APOSOR14391-1G APOSOR14391-250MG
tert-Butyl (piperazin-1-yl)acetate
tert-Butyl (piperazin-1-yl)acetate
Vzorec: C₁₀H₂₀N₂O₂
MW: 200.278 g/mol
Bod varu: 72 °C (0.15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06659535
CAS číslo: 112257-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR