1-Methyl-1H-imidazole-4-carbonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Cyano-1-methyl-1H-imidazole
OR14347-250MG
OR14347-1G
OR14347-5G
APOSOR14347-250MGEA
2780
CZK
APOSOR14347-250MG
APOSOR14347-1G
APOSOR14347-5G
1-Methyl-1H-imidazole-4-carbonitrile 97%
1-Methyl-1H-imidazole-4-carbonitrile
Vzorec:
C₅H₅N₃ Bod tání: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD06411537 CAS číslo: 66121-69-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...