Tisk…

3-Methylpyridine-2-acetamide oxime

Dodavatel: Apollo Scientific

Synonyms: N'-Hydroxy-3-methyl-2-pyridinecarboximidamide, N'-Hydroxy-3-methylpicolinimidamide

OR14020-1G OR14020-5G
APOSOR14020-1GEA 1210 CZK
APOSOR14020-1G APOSOR14020-5G
3-Methylpyridine-2-acetamide oxime
3-Methylpyridine-2-acetamide oxime
Vzorec: C₇H₉N₃O
MW: 151.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200877
CAS číslo: 690632-33-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR