Tisk…

5-Methylbenzo[b]thiophene-2-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR14014-1G OR14014-250MG
APOSOR14014-1GEA 10750 CZK
APOSOR14014-1G APOSOR14014-250MG
5-Methylbenzo[b]thiophene-2-sulphonyl chloride
5-Methylbenzo[b]thiophene-2-sulphonyl chloride
Vzorec: C₉H₇ClO₂S₂
MW: 246,734 g/mol
Bod tání: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD06200880
CAS číslo: 90273-30-6
UN: 2923
ADR: 8+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR