Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate 97%
Dodavatel: Apollo Scientific
OR13735-10G
OR13735-25G
OR13735-5G
APOSOR13735-10GEA
5200
CZK
APOSOR13735-10G
APOSOR13735-25G
APOSOR13735-5G
Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate 97%
Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate
Vzorec:
C₅H₇N₃O₂S MW: 173,193 g/mol Bod tání: 197…199 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03093787 CAS číslo: 64837-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...