Tisk…

3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulphonyl chloride, tech

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-[3-(Chlorosulphonyl)phenyl]-5-methyl-1,2,4-oxadiazole

OR1362-1G OR1362-250MG
APOSOR1362-1GEA 4960 CZK
APOSOR1362-1G APOSOR1362-250MG
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulphonyl chloride, tech
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulphonyl chloride
Vzorec: C₉H₇ClN₂O₃S
Bod tání: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD06797486
CAS číslo: 10185-62-3
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR