Tisk…

3-(Bromoacetyl)-4-methyl-1,2,5-oxadiazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Bromoacetyl)-4-methylfurazan, 2-Bromo-1-(4-methyl-1,2,5-oxadiazol-3-yl)ethan-1-one

OR13483-1G OR13483-100MG OR13483-250MG
APOSOR13483-1GEA 17160 CZK
APOSOR13483-1G APOSOR13483-100MG APOSOR13483-250MG
3-(Bromoacetyl)-4-methyl-1,2,5-oxadiazole
3-(Bromoacetyl)-4-methyl-1,2,5-oxadiazole
Vzorec: C₅H₅BrN₂O₂
MW: 205,009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00505140
CAS číslo: 165066-97-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR