N-Methyl-4-nitrobenzylamine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR13180-1G
OR13180-250MG
APOSOR13180-1GEA
744
CZK
APOSOR13180-1G
APOSOR13180-250MG
N-Methyl-4-nitrobenzylamine hydrochloride
N-Methyl-4-nitrobenzylamine hydrochloride
Vzorec:
C₈H₁₀N₂O₂·ClH MW: 202.638 g/mol Bod tání: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD06407995 CAS číslo: 19499-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...