Tisk…

Cyclopentylmethanesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR12996-1G OR12996-250MG OR12996-5G
APOSOR12996-1GEA 8570 CZK
APOSOR12996-1G APOSOR12996-250MG APOSOR12996-5G
Cyclopentylmethanesulphonyl chloride
Cyclopentylmethanesulphonyl chloride
Vzorec: C₆H₁₁ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD05863880
CAS číslo: 242459-85-4
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR