Tisk…

2-(Pyridin-3-yl)piperazine trihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Piperazin-2-yl)pyridine trihydrochloride

OR12885-250MG OR12885-1G
APOSOR12885-250MGEA 12200 CZK
APOSOR12885-250MG APOSOR12885-1G
2-(Pyridin-3-yl)piperazine trihydrochloride
2-(Pyridin-3-yl)piperazine trihydrochloride
Vzorec: C₉H₁₃N₃·3ClH
MW: 272,602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09702426
CAS číslo: 1171887-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR