Tisk…

2-Phenylpiperazine dihydrochloride

Dodavatel: Apollo Scientific

OR12883-1G OR12883-5G
APOSOR12883-1GEA 2240 CZK
APOSOR12883-1G APOSOR12883-5G
2-Phenylpiperazine dihydrochloride
2-Phenylpiperazine dihydrochloride
Vzorec: C₁₀H₁₄N₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD03840168
CAS číslo: 872139-22-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR