3-[(Piperidin-4-yl)methyl]pyridine dihydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-[(Pyridin-3-yl)methyl]piperidine dihydrochloride
OR12879-1G
OR12879-250MG
APOSOR12879-1GEA
32340
CZK
APOSOR12879-1G
APOSOR12879-250MG
3-[(Piperidin-4-yl)methyl]pyridine dihydrochloride 95%
3-[(Piperidin-4-yl)methyl]pyridine dihydrochloride
Vzorec:
C₁₁H₁₆N₂·2ClH MW: 249,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03840135 CAS číslo: 1172053-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...