Tisk…

N1,N1-Dimethyl-2-phenylethane-1,2-diamine

Dodavatel: Apollo Scientific
Varování

Synonyms: β-Amino-N,N-dimethylphenethylamine

OR12706-1G OR12706-250MG
APOSOR12706-1GEA 3580 CZK
APOSOR12706-1G APOSOR12706-250MG
N1,N1-Dimethyl-2-phenylethane-1,2-diamine
N1,N1-Dimethyl-2-phenylethane-1,2-diamine
Vzorec: C₁₀H₁₆N₂
MW: 164,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862528
CAS číslo: 31788-88-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR