Tisk…

4-(2-Chlorophenyl)benzylamine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: [4-(2-Chlorophenyl)phenyl]methylamine hydrochloride

OR12640-1G OR12640-250MG OR12640-50MG
APOSOR12640-1GEA 13230 CZK
APOSOR12640-1G APOSOR12640-250MG APOSOR12640-50MG
4-(2-Chlorophenyl)benzylamine hydrochloride
4-(2-Chlorophenyl)benzylamine hydrochloride
Vzorec: C₁₃H₁₂ClN·ClH
MW: 254,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02089431
CAS číslo: 518357-42-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR