Tisk…

N-Methyl-4-phenylbutylamine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: [4-(Methylamino)but-1-yl]benzene hydrochloride, 1-(Methylamino)-4-phenylbutane hydrochloride

OR12593-1G OR12593-5G OR12593-10G
APOSOR12593-1GEA 4530 CZK
APOSOR12593-1G APOSOR12593-5G APOSOR12593-10G
N-Methyl-4-phenylbutylamine hydrochloride
N-Methyl-4-phenylbutylamine hydrochloride
Vzorec: C₁₁H₁₇N·ClH
MW: 199,72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445574
CAS číslo: 148252-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR