Tisk…

3-(4-Methylphenoxy)benzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(Chlorosulphonyl)-4'-methyldiphenyl ether, 4-[3-(Chlorosulphonyl)phenoxy]toluene

OR12564-100MG OR12564-250MG
APOSOR12564-100MGEA 8600 CZK
APOSOR12564-100MG APOSOR12564-250MG
3-(4-Methylphenoxy)benzenesulphonyl chloride
3-(4-Methylphenoxy)benzenesulphonyl chloride
Vzorec: C₁₃H₁₁ClO₃S
MW: 282,743 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02089489
CAS číslo: 885950-86-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR